Learn More
MSC CM Transport Chain Assembly, 5/16 in, 4,700 Lb Work Load Limit, Grade 70, 20 ft
Catalog No. 5000510277
Encompass
This welded transport chain assembly, sized at 5/16 inch with clevis hooks, offers a robust 4,700 Lb work load limit. It is designed for secure and efficient transport applications, featuring a durable yellow chromate and chrome finish and a 20 foot length.
  • 4,700 Lb work load limit for heavy-duty applications.
  • Grade 70 chain ensures high strength and reliability.
  • 20 ft length provides ample reach for various transport needs.
  • Yellow chromate and chrome finish enhances durability and corrosion resistance.
  • Meets ASTM B633 Fe/ZN 13 Type II and Department of Transportation (D.O.T.) standards.
  • Manufactured in the United States.
Catalog No. 50-005-10277 Supplier MSC Supplier No. 87670907
This welded transport chain assembly, sized at 5/16 inch with clevis hooks, offers a robust 4,700 Lb work load limit. It is designed for secure and efficient transport applications, featuring a durable yellow chromate and chrome finish and a 20 foot length.
  • 4,700 Lb work load limit for heavy-duty applications.
  • Grade 70 chain ensures high strength and reliability.
  • 20 ft length provides ample reach for various transport needs.
  • Yellow chromate and chrome finish enhances durability and corrosion resistance.
  • Meets ASTM B633 Fe/ZN 13 Type II and Department of Transportation (D.O.T.) standards.
  • Manufactured in the United States.