Learn More
MSC CM Trolley, 1 Ton, Low Headroom, Ball Bearing
Catalog No. 5000516247
Encompass
This low headroom ball bearing trolley is designed for efficient material handling in various industrial settings. With a 1 ton (2000 lb) capacity, it is made of high-quality rolled steel and features a unique double row ball bearing wheel design for enhanced wear capacity and smooth operation.
  • Suitable for low headroom applications.
  • Negotiates radius curves as tight as 7 inches.
  • Double row, ball bearing wheel design for greater wear capacity.
  • Bearings are prepacked with lifetime lubricant.
  • Exclusive Dial-Fit collar adapts to a wide range of beams and patented rails.
  • Dust covers shield and protect bearings.
  • Adjustable for standard S-beams with depth of 4 to 12 inches.
  • Flange width adjustment from 2-5/8 to 5-1/16 inches.
  • Made in U.S.A.
Catalog No. 50-005-16247 Supplier MSC Supplier No. 89817712
This low headroom ball bearing trolley is designed for efficient material handling in various industrial settings. With a 1 ton (2000 lb) capacity, it is made of high-quality rolled steel and features a unique double row ball bearing wheel design for enhanced wear capacity and smooth operation.
  • Suitable for low headroom applications.
  • Negotiates radius curves as tight as 7 inches.
  • Double row, ball bearing wheel design for greater wear capacity.
  • Bearings are prepacked with lifetime lubricant.
  • Exclusive Dial-Fit collar adapts to a wide range of beams and patented rails.
  • Dust covers shield and protect bearings.
  • Adjustable for standard S-beams with depth of 4 to 12 inches.
  • Flange width adjustment from 2-5/8 to 5-1/16 inches.
  • Made in U.S.A.