Learn More
MSC CM Valustar Electric Chain Hoist, 1/4 Ton, 115V, 10 ft. Lift, 1 ea
Catalog No. 5000515724
Encompass
The Valustar Electric Chain Hoist is a compact and lightweight lifting solution engineered for general commercial applications. This unit offers a robust 1/4 ton (500 lb.) load capacity and provides dependable performance in a variety of industrial environments.
  • Features a heavy-duty caliper type AC brake for reliable stopping power.
  • Includes an efficient regenerative braking system to minimize heat generation.
  • Equipped with a fully machined and heat-treated liftwheel with hardened chain guides.
  • Utilizes a 115V power supply and rugged NEMA 4 control station.
  • Built with a 1/4 HP motor and single reeving configuration.
  • Standard overload protector device provided for enhanced safety.
  • Gear train is lifetime lubricated with non-oxidizing grease.
  • Constructed with latch-type hooks and hook suspension.
Catalog No. 50-005-15724 Supplier MSC Supplier No. 89825400
The Valustar Electric Chain Hoist is a compact and lightweight lifting solution engineered for general commercial applications. This unit offers a robust 1/4 ton (500 lb.) load capacity and provides dependable performance in a variety of industrial environments.
  • Features a heavy-duty caliper type AC brake for reliable stopping power.
  • Includes an efficient regenerative braking system to minimize heat generation.
  • Equipped with a fully machined and heat-treated liftwheel with hardened chain guides.
  • Utilizes a 115V power supply and rugged NEMA 4 control station.
  • Built with a 1/4 HP motor and single reeving configuration.
  • Standard overload protector device provided for enhanced safety.
  • Gear train is lifetime lubricated with non-oxidizing grease.
  • Constructed with latch-type hooks and hook suspension.