Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ CM6010 Hard Hat Mounted Ear Muff

Catalog No. 19155293 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
27 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Noise Reduction Rating (NRR)
19-155-293 27 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-155-293 Supplier Pyramex™ Supplier No. CM6010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CM6010 fits a wide variety of hard hats and ear muff attachments can be easily flipped up or down when needed.

  • Fits a wide variety of hard hats
  • Dielectric
  • Soft foam ear cups
  • Easily accessible while working
  • Cups flip up or down over ears depending on timing of use

Specifications

Color Gray
ANSI S3.1974 Yes
Cushion Material PVC
Cushion Color Black
For Use With (Equipment) Wide variety of hard hats
Model CM6010
Packaging Type Poly bag
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 27 dB
Dielectric Yes
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.