Learn More
Aviva Systems Biology Corp CMAS Antibody - N-terminal region (ARP48845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CMAS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CMAS. Alternate names: CSSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59937