Learn More
MSC Proto Combination Wrench, 32.00 mm, Satin Finish, 1 ea
Catalog No. 501920469
Encompass
The Proto combination wrench is engineered for reliable performance in demanding environments. Featuring a 32.00 mm head size, this steel wrench with a satin finish offers durability and a 15-degree offset for improved access in tight spaces.
  • Manufactured from steel for strength and longevity
  • Satin finish provides corrosion resistance and a comfortable grip
  • Features a 32.00 mm head size for metric fasteners
  • Provides 15 degree head offset for enhanced clearance
  • Tether capable for secure working at height
  • Corrosion-resistant for extended tool life
  • Designed with 12 points for efficient torque application
  • Meets ASME B107.100 standards for quality and safety
  • Backed by a lifetime warranty for assured performance
Catalog No. 50-192-0469 Supplier MSC Supplier No. 38735304
The Proto combination wrench is engineered for reliable performance in demanding environments. Featuring a 32.00 mm head size, this steel wrench with a satin finish offers durability and a 15-degree offset for improved access in tight spaces.
  • Manufactured from steel for strength and longevity
  • Satin finish provides corrosion resistance and a comfortable grip
  • Features a 32.00 mm head size for metric fasteners
  • Provides 15 degree head offset for enhanced clearance
  • Tether capable for secure working at height
  • Corrosion-resistant for extended tool life
  • Designed with 12 points for efficient torque application
  • Meets ASME B107.100 standards for quality and safety
  • Backed by a lifetime warranty for assured performance