Learn More
MSC Proto Combination Wrench, 1-5/16 in., 15 deg Offset, 1 ea
Catalog No. 501920489
Encompass
This combination wrench features a 1-5/16 in. head size and a 15-degree offset. Constructed from durable steel, it is designed for demanding applications requiring both open-end and box-end functionality.
  • Head size of 1-5/16 in.
  • Features a 15-degree offset head
  • Straight handle design
  • Durable steel construction
  • Overall length of 17-5/8 in.
  • Tether capable for safety
  • Corrosion-resistant finish
  • Compliant with ASME B107.100-2010 standards
  • Provides 12 points of contact for secure grip
Catalog No. 50-192-0489 Supplier MSC Supplier No. 88468459
This combination wrench features a 1-5/16 in. head size and a 15-degree offset. Constructed from durable steel, it is designed for demanding applications requiring both open-end and box-end functionality.
  • Head size of 1-5/16 in.
  • Features a 15-degree offset head
  • Straight handle design
  • Durable steel construction
  • Overall length of 17-5/8 in.
  • Tether capable for safety
  • Corrosion-resistant finish
  • Compliant with ASME B107.100-2010 standards
  • Provides 12 points of contact for secure grip