Learn More
MSC Proto Combination Wrench, SAE, 15/16 in, 15 deg, 1 ea
Catalog No. 501920107
Encompass
This combination wrench from Proto is designed for efficient fastening and helps prevent fastener rounding. It features a full polish chrome finish for enhanced corrosion resistance and easy cleaning, suitable for various professional applications.
  • Provides corrosion resistance and easy-to-clean surfaces
  • Six-point box end ensures large wrench-to-fastener contact to prevent rounding
  • Anti-Slip Design (ASD) open end for secure grip (excludes fractional sizes 11/32 in and smaller)
  • Box ends incorporate TorquePlus™ system for improved torque transmission
  • Tether capable for secure usage at heights or in critical environments
  • Constructed from durable alloy steel
  • Meets ASME B107.100-2010 standards
Catalog No. 50-192-0107 Supplier MSC Supplier No. 38734521
This combination wrench from Proto is designed for efficient fastening and helps prevent fastener rounding. It features a full polish chrome finish for enhanced corrosion resistance and easy cleaning, suitable for various professional applications.
  • Provides corrosion resistance and easy-to-clean surfaces
  • Six-point box end ensures large wrench-to-fastener contact to prevent rounding
  • Anti-Slip Design (ASD) open end for secure grip (excludes fractional sizes 11/32 in and smaller)
  • Box ends incorporate TorquePlus™ system for improved torque transmission
  • Tether capable for secure usage at heights or in critical environments
  • Constructed from durable alloy steel
  • Meets ASME B107.100-2010 standards