Learn More
MSC Proto Combination Wrench, 16 mm, 15 deg Offset, 1 ea
Catalog No. 501920460
Encompass
This Proto combination wrench is designed for mechanical applications, offering efficient fastening and loosening of metric fasteners. It features a durable steel construction with a satin finish, suitable for demanding work environments.
  • Features a 16 mm head size
  • Has a satin finish
  • Offset head type for improved access
  • Straight handle for comfortable grip
  • Constructed from durable steel material
  • Corrosion-resistant for extended lifespan
  • Complies with ASME B107.100 standards
  • Backed by a lifetime warranty
  • Equipped with 12 points of contact
  • Designed with a 15 degree head offset angle
  • Offers a 15 degree opening angle
Catalog No. 50-192-0460 Supplier MSC Supplier No. 38735296
This Proto combination wrench is designed for mechanical applications, offering efficient fastening and loosening of metric fasteners. It features a durable steel construction with a satin finish, suitable for demanding work environments.
  • Features a 16 mm head size
  • Has a satin finish
  • Offset head type for improved access
  • Straight handle for comfortable grip
  • Constructed from durable steel material
  • Corrosion-resistant for extended lifespan
  • Complies with ASME B107.100 standards
  • Backed by a lifetime warranty
  • Equipped with 12 points of contact
  • Designed with a 15 degree head offset angle
  • Offers a 15 degree opening angle