Learn More
MSC Proto Combination Wrench, 24.00 mm, 15 deg Offset, 1 ea
Catalog No. 501920464
Encompass
This combination wrench is designed for professional use, featuring a 24.00 mm head size and a 15-degree offset for improved access. Constructed from steel with a satin finish, it offers durability and corrosion resistance for demanding applications.
  • Features a 24.00 mm head size
  • Offers a 15 degree offset head
  • Constructed from durable steel
  • Satin finish for corrosion resistance
  • Tether capable design
  • Straight handle type
  • Meets ASME B107.100 standards
  • Lifetime warranty
  • Equipped with 12 points for secure grip
Catalog No. 50-192-0464 Supplier MSC Supplier No. 38735064
This combination wrench is designed for professional use, featuring a 24.00 mm head size and a 15-degree offset for improved access. Constructed from steel with a satin finish, it offers durability and corrosion resistance for demanding applications.
  • Features a 24.00 mm head size
  • Offers a 15 degree offset head
  • Constructed from durable steel
  • Satin finish for corrosion resistance
  • Tether capable design
  • Straight handle type
  • Meets ASME B107.100 standards
  • Lifetime warranty
  • Equipped with 12 points for secure grip