Learn More
MSC Proto Combination Wrench, 9/16 in., 12-Point, Short, 1 ea
Catalog No. 501920131
Encompass
Proto combination wrenches are designed for efficient fastening in various applications. This 9/16 in. full polish 12-point short combination wrench offers enhanced access and durability for professional use.
  • Twelve point box end allows for fine indexing, helping to improve speed and access. Fits 4, 6 and 12 point fasteners.
  • The wrench self aligns with the fastener.
  • Full polish finish provides corrosion resistance and easy to clean surfaces.
  • The box end is countersunk for easy application to the fastener.
  • Fifteenº angled offset helps keep shank clear of obstructions.
  • Anti-slip design (ASD) open end.
  • Box ends incorporate the TorquePlus™ system.
Catalog No. 50-192-0131 Supplier MSC Supplier No. 38855342
Proto combination wrenches are designed for efficient fastening in various applications. This 9/16 in. full polish 12-point short combination wrench offers enhanced access and durability for professional use.
  • Twelve point box end allows for fine indexing, helping to improve speed and access. Fits 4, 6 and 12 point fasteners.
  • The wrench self aligns with the fastener.
  • Full polish finish provides corrosion resistance and easy to clean surfaces.
  • The box end is countersunk for easy application to the fastener.
  • Fifteenº angled offset helps keep shank clear of obstructions.
  • Anti-slip design (ASD) open end.
  • Box ends incorporate the TorquePlus™ system.