Learn More
Selleck Chemical LLC CMC-Na
SDP
Catalog No. 5000002764
Encompass_Preferred
CMC-Na (Sodium carboxymethyl cellulose) is one of the important modified cellulose a water-soluble cellulose which is widely used in many application of food pharmaceuticals detergent paper coating dispersing agent and others Its addition possibly increases the hydrogenation and dehydrogenation features of Magnesium
Catalog No. 50-000-02764 Supplier Selleck Chemical LLC Supplier No. S670310G
CMC-Na (Sodium carboxymethyl cellulose) is one of the important modified cellulose a water-soluble cellulose which is widely used in many application of food pharmaceuticals detergent paper coating dispersing agent and others Its addition possibly increases the hydrogenation and dehydrogenation features of Magnesium