Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMC Rescue™ Shrink Tubing

Catalog No. 19822075
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-075 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-075 Supplier CMC Rescue™ Supplier No. 294358
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Clear tubing protects labels on the ends of ropes

  • Slide tubing over label
  • Use safe heat source to shrink tubing securely to rope

Specifications

Color Clear
For Use With (Equipment) 0.5, 0.6 in. dia. Rope
Product Type Tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.