Learn More
Medchemexpress LLC HY-120793 5mg Medchemexpress, CMC2.24 CAS:1255639-43-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1207935MG
Encompass_Preferred
Medchemexpress, HY-120793 5mg CMC2.24 CAS:1255639-43-0 CMC2.24 (TRB-N0224), an orally active tricarbonylmethane agent, is effective against pancreatic tumor in mice by inhibiting Ras activation and its downstream effector ERK1/2 pathway. CMC2.24 is also a potent inhibitor of zinc-dependent MMPs with IC50s ranging from 2.0-69 μM. CMC2.24 alleviates osteoarthritis progression by restoring cartilage homeostasis and inhibiting chondrocyte apoptosis via the NF-κB/HIF-2α axis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0952