Learn More
Electron Microscopy Sciences CMCP-9 Low viscosity Mountant 500 ML
SDP
Supplier:  Electron Microscopy Sciences 1800305
Encompass_Preferred
Colorless, non resinous, water miscible mounting medium is used like Canada Balsam to mount almost any material, provided it is not stained, calciferous material, or thin tissue sections. This permanent transparent mounting media has the advantage of allowing live or preserved specimens to be mounted directly from water or alcohol. Free living nematodes and live parasitic worms are among the organism suitable for mounting with CMCP. The CMCP mounting provide then with rapid relaxation, death, and clearing action.
Catalog No. 50-190-0429