Learn More
Medchemexpress LLC HY-P1453A 1mg Medchemexpress, CMD178 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1453A1MG
Encompass_Preferred
Medchemexpress, HY-P1453A 1mg CMD178 (TFA) CAS: CMD178 (TFA) is a lead peptide that consistently reduces the expression of Foxp3 and STAT5 induced by IL-2/s IL-2Rα signaling. CMD178 (TFA) also is an inhibitor of STAT5 and inhibits T reg cells development [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4203