Learn More
Medchemexpress LLC CMD178 TFA | 99.7% | 964.05 | 5 MG
SDP
Catalog No. 5000424743
Encompass_Preferred
CMD178 (TFA) is a lead peptide that consistently reduces the expression of Foxp3 and STAT5 induced by IL-2/s IL-2Ra signaling. It also acts as an inhibitor of STAT5 and inhibits Treg cells development.
  • Reduces expression of Foxp3 and STAT5 induced by IL-2/s IL-2Ra signaling
  • Inhibits STAT5
  • Inhibits Treg cells development
Catalog No. 50-004-24743 Supplier Medchemexpress LLC Supplier No. HYP1453A5MG
CMD178 (TFA) is a lead peptide that consistently reduces the expression of Foxp3 and STAT5 induced by IL-2/s IL-2Ra signaling. It also acts as an inhibitor of STAT5 and inhibits Treg cells development.
  • Reduces expression of Foxp3 and STAT5 induced by IL-2/s IL-2Ra signaling
  • Inhibits STAT5
  • Inhibits Treg cells development