Learn More
Echelon Biosciences Research Labs 2-C-Methyl-D-erythritol 2,4-cyclophosphate (cMEPP) 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs IM0541MG
Encompass
2-C-Methyl-D-erythritol 2,4-cyclophosphate (cMEPP) is an intermediate in the non-mevalonate pathway. It is formed by from MECDP Synthase from CDP-MEP and converted to 1-hydroxy-2-methyl-2-buten-4-yl diphosphate (HDMAPP) by IspG (GcpE). This item is non returnable
Catalog No. 50-720-404