Learn More
Sino Biological c-Met / HGFR Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 10692R271100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human c-Met / HGFR (rh c-Met / HGFR; Catalog#10692-H08H; NP_000236.2; Met1-Thr932).
Catalog No. 50-224-7360