Learn More
Enzo Life Sciences CMH. hydrochloride (250mg)
SDP
Supplier:  Enzo Life Sciences ALX430117M250
Encompass_Preferred
Analog of CPH (ALX-430-078) with higher cell permeability and 3-fold better reactivity with superoxide. Has been used to study intracellular production of superoxide radicals in plasma, endothelial cells and isolated hearts. Alternative name: 1-Hydroxy-3-methoxycarbonyl-2,2,5,5-tetramethylpyrrolidine . HCl. Purity: ≥99% (EPR). Solubility: Soluble in DMSO, 100% ethanol, methanol or water. Long Term Storage: -20°C.
Catalog No. 50-200-9209