Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMIT/MIT Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002598 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2598 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2598 Supplier MilliporeSigma Supelco Supplier No. PHR15971ML
Add to Cart
Add to Cart
Pharmaceutical secondary standard for applications in quality control, provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.CMIT/MIT is an isothiazolone biocide having a 3:1 ratio of CMIT and MIT, widely used for its broad-spectrum action against microbes, algae, and fungi. It is one of the active ingredients of humidifier disinfectants and a commonly used preservative in industrial products such as cosmetics, paints, adhesives and detergents. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 26172-55-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Synonym 5-chloro-2-methyl-2 h-isothiazolin-3-one/2-methyl-2 h-isothiazol-3-one
Type Certified Reference Material
Quantity 1 mL
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.