Learn More
ABclonal Technology CMKLR1 Rabbit pAb
Supplier: ABclonal Technology A15237500UL
Enables adipokinetic hormone binding activity and adipokinetic hormone receptor activity. Involved in several processes, including negative regulation of NF-kappaB transcription factor activity, positive regulation of macrophage chemotaxis, and regulation of calcium-mediated signaling. Located in plasma membrane.