Learn More
Medchemexpress LLC HY-124828 5mg Medchemexpress, CMLD-2 CAS:958843-91-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1248285MG
Encompass_Preferred
Medchemexpress, HY-124828 5mg CMLD-2 CAS:958843-91-9 CMLD-2, an inhibitor of HuR-ARE interaction, competitively binds HuR protein disrupting its interaction with adenine-uridine rich elements (ARE)-containing mRNAs (Ki=350 nM). CMLD-2 induces apoptosis exhibits antitumor activity in different cancer cells as colon, pancreatic, thyroid and lung cancer cell lines. Hu antigen R (HuR) is an RNA binding protein, can regulate target mRNAs stability and translation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1056