Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMNB-Caged Carboxyfluorescein, SE (5-Carboxyfluorescein-Bis-(5-Carboxymethoxy-2-Nitrobenzyl) Ether, β-Alanine-Carboxamide, Succinimidyl Ester)

Catalog No. C20050
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20050 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20050 Supplier Invitrogen™ Supplier No. C20050
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

CMNB-Caged Carboxyfluorescein, SE (5-Carboxyfluorescein-Bis-(5-Carboxymethoxy-2-Nitrobenzyl) Ether, β-Alanine-Carboxamide, Succinimidyl Ester)

Conjugation of the succinimidyl ester of the water-soluble, CMNB-caged carboxyfluorescein, SE to a primary amine on a biomolecule of interest produces an essentially nonfluorescent probe that yields a green-fluorescent fluorescein-labeled product only after ultraviolet illumination and removal of the caging group.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type CMNB-Caged Carboxyfluorescein
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.