Learn More
Medchemexpress LLC CMP-Sialic acid sodium salt | 1007117-62-5 | 90.8% | 636.43 | 1 MG
SDP
Catalog No. 5000439752
Encompass_Preferred
CMP-Sialic acid (CMP-Neu5Ac) sodium salt is an allosteric inhibitor of UDP-GlcNAc 2-epimerase. It provides a substrate for Golgi sialyltransferases and is an important sugar nucleotide for the biosynthesis of sialic acid and its conjugates.
  • Functions as an allosteric inhibitor of UDP-GlcNAc 2-epimerase.
  • Serves as a substrate for Golgi sialyltransferases.
  • Important sugar nucleotide for biosynthesis of sialic acid and its conjugates.
  • Can be used as a CMAH substrate to detect Neu5Gc formation in platelet lysates.
Catalog No. 50-004-39752 Supplier Medchemexpress LLC Supplier No. HY112942A1MG
CMP-Sialic acid (CMP-Neu5Ac) sodium salt is an allosteric inhibitor of UDP-GlcNAc 2-epimerase. It provides a substrate for Golgi sialyltransferases and is an important sugar nucleotide for the biosynthesis of sialic acid and its conjugates.
  • Functions as an allosteric inhibitor of UDP-GlcNAc 2-epimerase.
  • Serves as a substrate for Golgi sialyltransferases.
  • Important sugar nucleotide for biosynthesis of sialic acid and its conjugates.
  • Can be used as a CMAH substrate to detect Neu5Gc formation in platelet lysates.