Learn More
Medchemexpress LLC CMP-sialic acid sodium salt | 1007117-62-5 | MFCD00151579 | 90.8% | 636.43 g/mol | C20H30N4NaO16P | 5 MG
SDP
Catalog No. 5000264014
Encompass_Preferred
CMP-Sialic acid sodium salt is a nucleotide sugar involved in the biosynthesis of sialic acid. It functions as an allosteric inhibitor of UDP-GlcNAc 2-epimerase and provides a substrate for Golgi sialyltransferases, making it useful for enzymatic sialylation and studies of glycan modification in biochemical research.
  • Provides substrate activity for Golgi sialyltransferases.
  • Acts as an allosteric inhibitor of UDP-GlcNAc 2-epimerase.
  • Supports enzymatic sialylation and glycan modification studies.
  • Supplied as a solid, white to off-white, with defined storage stability.
  • Available in small research-scale quantities for laboratory use.
Catalog No. 50-002-64014 Supplier Medchemexpress LLC Supplier No. HY112942A5MG
CMP-Sialic acid sodium salt is a nucleotide sugar involved in the biosynthesis of sialic acid. It functions as an allosteric inhibitor of UDP-GlcNAc 2-epimerase and provides a substrate for Golgi sialyltransferases, making it useful for enzymatic sialylation and studies of glycan modification in biochemical research.
  • Provides substrate activity for Golgi sialyltransferases.
  • Acts as an allosteric inhibitor of UDP-GlcNAc 2-epimerase.
  • Supports enzymatic sialylation and glycan modification studies.
  • Supplied as a solid, white to off-white, with defined storage stability.
  • Available in small research-scale quantities for laboratory use.