Learn More
TARGETMOL CHEMICALS INC CMP-Sialic acid sodium salt
SDP
Catalog No. 502880789
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. CMP-Sialic acid sodium salt (CMP-Neu5Ac sodium salt) is a variant inhibitor of UDP-GlcNAc 2-epimerase, the activated form of sialic acid, which is widely found in animals and is involved in the metabolism of organisms. Purity 98.32%
Catalog No. 50-288-0789 Supplier TARGETMOL CHEMICALS INC Supplier No. T108512MG
Also available in 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. CMP-Sialic acid sodium salt (CMP-Neu5Ac sodium salt) is a variant inhibitor of UDP-GlcNAc 2-epimerase, the activated form of sialic acid, which is widely found in animals and is involved in the metabolism of organisms. Purity 98.32%