Learn More
Cayman Chemical CMP-SIalIc AcIdodIum salt 10mg
SDP
Catalog No. 502970564
Encompass_Preferred
A form of the sugar Neu5Ac O-linked with the nucleotide CMP; serves as the substrate for sialyltransferases, which transfer Neu5Ac from CMP-sialic acid to various acceptor substrates, most commonly at terminal positions of the oligosaccharide component of glycoproteins or glycolipids
Catalog No. 50-297-0564 Supplier Cayman Chemical Supplier No. 1640410 MG
A form of the sugar Neu5Ac O-linked with the nucleotide CMP; serves as the substrate for sialyltransferases, which transfer Neu5Ac from CMP-sialic acid to various acceptor substrates, most commonly at terminal positions of the oligosaccharide component of glycoproteins or glycolipids