Learn More
McMaster-Carr CMP TOTE BOX 22.5L X17.5W X 8H
Catalog No. NC2587065
Encompass
When used with a lid, these tote boxes protect contents from electrostatic discharge.
Catalog No. NC2587065 Supplier McMaster-Carr Supplier No. 4893T29
When used with a lid, these tote boxes protect contents from electrostatic discharge.