Learn More
OTT Hydromet Kipp & Zonen - CMP3 PYRANOMETER
Supplier:  OTT Hydromet 0338920002
Encompass_Preferred
The CMP3 pyranometer is an instrument for measuring the solar irradiance. The thermopile sensor construction measures the solar energy received from the total solar spectrum and the whole hemisphere (180 degrees field of view). The output is expressed in Watts per meter square. The CMP3 pyranometer is designed for continuous indoor and outdoor use. The CMP3 pyranometer (ISO 9060 spectrally flat Class C) is intended for shortwave global solar radiation measurements in the spectral range from 300 to 2800 nm. The thermopile detector measures irradiance up to 2000 W/m² with response time <18 seconds and typical sensitivity 10 µV/W/m² that varies less than 5 % from -10 ºC to +40 ºC. Two CMP 3s can easily be mounted back-to-back to make a low-cost albedometer. The Pyranometer doesn’t require any power, it supplies a low voltage of 0 to 20mV in relation to the amount of incoming radiation. When a higher voltage level or a 4 to 20mA signal is required, the AMPBOX is the perfect solution.
Catalog No. NC1919645