Learn More
Hach Company Compact Analyzer Thumb Screw (stainless steel, 6/32") - Replacement for CL17 and SP510 analyzers
Catalog No. NC2590120 Shop All Hach Company Products
Encompass
Replacement washer-face thumb screw made from stainless steel with a 6/32" thread, used to secure pressure plates and washers on compact process analyzers. Fits CL17 and SP510 analyzers and provides corrosion-resistant fastening for reliable operation in laboratory and process environments.
  • Stainless steel construction for corrosion resistance.
  • 6/32" thread size for secure fastening.
  • Washer-face design for stable sealing and retention.
  • Replacement part to minimize analyzer downtime.
  • Compact form factor for easy installation in analyzer assemblies.
Catalog No. NC2590120 Supplier Hach Company Supplier No. 5410100
Replacement washer-face thumb screw made from stainless steel with a 6/32" thread, used to secure pressure plates and washers on compact process analyzers. Fits CL17 and SP510 analyzers and provides corrosion-resistant fastening for reliable operation in laboratory and process environments.
  • Stainless steel construction for corrosion resistance.
  • 6/32" thread size for secure fastening.
  • Washer-face design for stable sealing and retention.
  • Replacement part to minimize analyzer downtime.
  • Compact form factor for easy installation in analyzer assemblies.