Learn More
Medchemexpress LLC CMPD1 10MG
SDP
Catalog No. NC2544580
Encompass_Preferred
CMPD1 is a selective and non-ATP-competitive p38 MAPK-mediated MK2 phosphorylation inhibitor with apparent Ki (Kiapp) of 330 nM., CAS 41179-33-3
Catalog No. NC2544580 Supplier Medchemexpress LLC Supplier No. HY10864310MG
CMPD1 is a selective and non-ATP-competitive p38 MAPK-mediated MK2 phosphorylation inhibitor with apparent Ki (Kiapp) of 330 nM., CAS 41179-33-3