Learn More
Medchemexpress LLC CMPD1 | 41179-33-3 | 99.9% | 349.40 | 10 MG
SDP
Catalog No. 5000344297
Encompass_Preferred
CMPD1 is a selective and non-ATP-competitive p38 MAPK-mediated MK2 phosphorylation inhibitor with an apparent Ki (Kiapp) of 330 nM. It is intended for research use only and not for sale to patients.
  • Does not inhibit p38 MAPK-mediated phosphorylation of other substrates, MBP and ATF2.
  • Induces mitotic arrest and apoptosis in U87 cells.
  • Inhibits tubulin polymerisation in glioblastoma cells.
Catalog No. 50-003-44297 Supplier Medchemexpress LLC Supplier No. HY108643100MG
CMPD1 is a selective and non-ATP-competitive p38 MAPK-mediated MK2 phosphorylation inhibitor with an apparent Ki (Kiapp) of 330 nM. It is intended for research use only and not for sale to patients.
  • Does not inhibit p38 MAPK-mediated phosphorylation of other substrates, MBP and ATF2.
  • Induces mitotic arrest and apoptosis in U87 cells.
  • Inhibits tubulin polymerisation in glioblastoma cells.