Learn More
Medchemexpress LLC HY-108643 5mg Medchemexpress, CMPD1 CAS:41179-33-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086435MG
Encompass_Preferred
Medchemexpress, HY-108643 5mg CMPD1 CAS:41179-33-3 CMPD1 is a selective and non-ATP-competitive p38 MAPK-mediated MK2 phosphorylation inhibitor with apparent K i (K i app ) of 330 nM [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2327