Learn More
Medchemexpress LLC Cmpf-d5 | 2749807-07-4 | 99.0% | 1 MG
SDP
Catalog No. 5000339550
Encompass_Preferred
CMPF-d5 is the deuterium labeled CMPF, which serves as a valuable tool for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The strategic deuterium substitution in drug molecules has garnered attention due to its potential to significantly impact their pharmacokinetic and metabolic profiles, offering insights into drug development processes.
  • Functions as a tracer or internal standard in quantitative analytical methods.
  • Influences the pharmacokinetic and metabolic profiles of various drugs.
  • Recommended storage for powder is -20°C for up to 3 years.
  • Recommended storage for solutions is -80°C for 6 months or -20°C for 1 month.
  • Requires careful handling, avoiding inhalation and contact with skin or eyes.
  • Store in a tightly sealed container within a cool, well-ventilated environment.
Catalog No. 50-003-39550 Supplier Medchemexpress LLC Supplier No. HY129297S1MG
CMPF-d5 is the deuterium labeled CMPF, which serves as a valuable tool for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The strategic deuterium substitution in drug molecules has garnered attention due to its potential to significantly impact their pharmacokinetic and metabolic profiles, offering insights into drug development processes.
  • Functions as a tracer or internal standard in quantitative analytical methods.
  • Influences the pharmacokinetic and metabolic profiles of various drugs.
  • Recommended storage for powder is -20°C for up to 3 years.
  • Recommended storage for solutions is -80°C for 6 months or -20°C for 1 month.
  • Requires careful handling, avoiding inhalation and contact with skin or eyes.
  • Store in a tightly sealed container within a cool, well-ventilated environment.