Learn More
Med Vet International CMPK, 50 Bolus, 24gm each
Catalog No. 502188557
Encompass_Preferred
For use in fresh cows to help maintain normal levels of calcium, magnesium, phosphorus, and potassium around freshening. May also be used in horses, sheep and swine. Ingredients: Dicalcium phosphate, dextrose, magnesium oxide, sodium chloride, calcium carbonate, potassium chloride, dl-alpha-tocopherol acetate, corn starch, Vitamin D3 supplement, and magnesium stearate. Guaranteed Analysis (per 24 grams) - Calcium (min.), 13.0% - Calcium (max.), 15.6% - Phosphorus, 8.0% - Salt (min.), 5.0% - Salt (max.), 5.6% - Potassium, 4.0% - Magnesium, 6.0% - Vitamin D3, 50000 IU - Vitamin E, 300 IUProducts are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-218-8557 Supplier Med Vet International Supplier No. PHCMPK50
For use in fresh cows to help maintain normal levels of calcium, magnesium, phosphorus, and potassium around freshening. May also be used in horses, sheep and swine. Ingredients: Dicalcium phosphate, dextrose, magnesium oxide, sodium chloride, calcium carbonate, potassium chloride, dl-alpha-tocopherol acetate, corn starch, Vitamin D3 supplement, and magnesium stearate. Guaranteed Analysis (per 24 grams) - Calcium (min.), 13.0% - Calcium (max.), 15.6% - Phosphorus, 8.0% - Salt (min.), 5.0% - Salt (max.), 5.6% - Potassium, 4.0% - Magnesium, 6.0% - Vitamin D3, 50000 IU - Vitamin E, 300 IUProducts are not always manufacturer specific. Actual product appearance may differ from website image.