Learn More
Aviva Systems Biology Corp Cmpk1 Antibody - C-terminal region (ARP61819_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cmpk1. It was validated on Western Blot. . Alternate names: 0610011D08Rik, Cmpk, UMP-CMPk, RP23-295C20.4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-477