Learn More
Chemglass Life Sciences 3/4" PFA Elbow Compression Fitting
SDP
Catalog No. 50143655
Encompass_Preferred
3/4" PFA Elbow Compression Fitting. PFA elbow compression fittings are ideally suited for use in corrosive environments and chemical applications. The fitting design yields a leak-tight connection and requires no special tools to assemble. The connection is achieved by merely inserting the tubing into the fitting assembly and tightening the nut.
Catalog No. 50-143-655 Supplier Chemglass Life Sciences Supplier No. CG1975D24
3/4" PFA Elbow Compression Fitting. PFA elbow compression fittings are ideally suited for use in corrosive environments and chemical applications. The fitting design yields a leak-tight connection and requires no special tools to assemble. The connection is achieved by merely inserting the tubing into the fitting assembly and tightening the nut.