Learn More
Medchemexpress LLC HY-135981 5mg Medchemexpress, CMS-121 CAS:1353224-53-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1359815MG
Encompass_Preferred
Medchemexpress, HY-135981 5mg CMS-121 CAS:1353224-53-9 CMS-121 is a quinolone derivative and an orally active acetyl-CoA carboxylase 1 (ACC1) inhibitor. CMS-121 protects HT22 cells against ischemia and oxidative damage with EC50 values of 7 nM and 200 nM, respectively. CMS-121 has strong neuroprotective, anti-inflammatory, antioxidative and renoprotective activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1416