Learn More
ATCC CMT-93; Rectal Carcinoma; Mouse (Mus musculus)
Catalog No. 502384748
Encompass
CMT-93 is a cell line exhibiting epithelial morphology that was isolated from the rectum of a mouse with polyploid carcinoma. This cell line was deposited by LM Franks and can be used in cancer research.
Catalog No. 50-238-4748 Supplier ATCC Supplier No. CCL223
CMT-93 is a cell line exhibiting epithelial morphology that was isolated from the rectum of a mouse with polyploid carcinoma. This cell line was deposited by LM Franks and can be used in cancer research.