Learn More
Aviva Systems Biology Corp CMTM8 Antibody - middle region (ARP53386_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53386_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CMTM8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CMTM8. Alternate names: CKLFSF8, CKLFSF8-V2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61013