Learn More
BIOHIPPO CMV-ACVR1C-RFP-BSD
SDP
Catalog No. 5000331121
Encompass
CMV-ACVR1C-RFP-BSD lentiviral vector expressing RFP under CMV promoter Each vial provides a nominal titer of 2x10E6 transducing units TU Human ACVR2B ORF cDNA Lentivirus Lentiviral particles packaged with expression vector for human ACVR2B (uniprot ID Q8NER5) A CMV promoter drives expression of the ORF fused to a C-terminal epitope tag and fluorescent reporter which is separated by a self-cleaving peptide These lentiviral particles allow for easy transduction and high stable expression of the indicated ORF cDNA in mammalian cells The lentiviral vectors have been optimized to provide high gene expression and reliable genome integration allowing for easy establishment of long-term stable cell lines with consistent read out of fluorescent (GFP RFP BFP) or luminescent (firefly luc) reporters
Catalog No. 50-003-31121 Supplier BIOHIPPO Supplier No. BHV194001183
CMV-ACVR1C-RFP-BSD lentiviral vector expressing RFP under CMV promoter Each vial provides a nominal titer of 2x10E6 transducing units TU Human ACVR2B ORF cDNA Lentivirus Lentiviral particles packaged with expression vector for human ACVR2B (uniprot ID Q8NER5) A CMV promoter drives expression of the ORF fused to a C-terminal epitope tag and fluorescent reporter which is separated by a self-cleaving peptide These lentiviral particles allow for easy transduction and high stable expression of the indicated ORF cDNA in mammalian cells The lentiviral vectors have been optimized to provide high gene expression and reliable genome integration allowing for easy establishment of long-term stable cell lines with consistent read out of fluorescent (GFP RFP BFP) or luminescent (firefly luc) reporters