Learn More
Cellecta Inc Lentiviral CMV-Cas9-Hyg Expression Vector, packaged virus, 1 x 10^6 TU

Supplier: Cellecta Inc SVC9VS
This Cellecta CMV-Cas9-Hyg lentiviral expression construct is designed for stable Cas9 expression across various cell types, promoting efficient CRISPR-mediated gene editing and screening applications. With a robust CMV promoter and a hygromycin resistance gene, it enables effective selection of transduced cells. Packaged as a lentivirus, it excels in high-efficiency transduction in both dividing and non-dividing mammalian cells.
- Promoter used for Cas9 expression is CMV
- Cas9 origin is *S. pyogenes*
- Hygromycin resistance gene for selection
- Compatible with dividing and non-dividing cells
- Stored at -80°C
- Has a shelf life of 1 year from receipt
- Offers a quantity of 1 x 10^6 TU
- Application includes gene editing and genetic screening
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.