Learn More
Newcomer Supply Cytomegalovirus (CMV) Control Slides, 98/set
SDP
Catalog No. NC0568445
Encompass
Newcomer Supply Cytomegalovirus (CMV) Control Slides are for the positive immunohistochemical staining of cytomegalovirus, a member of the herpes virus group, including herpes simplex virus types 1 and 2, varicella zoster virus, and Epstein Barr virus. Cytomegalovirus (CMV) Control Slides contain sections of positive staining lung and negative staining myometrium.
Catalog No. NC0568445 Supplier Newcomer Supply Supplier No. 3240B
Newcomer Supply Cytomegalovirus (CMV) Control Slides are for the positive immunohistochemical staining of cytomegalovirus, a member of the herpes virus group, including herpes simplex virus types 1 and 2, varicella zoster virus, and Epstein Barr virus. Cytomegalovirus (CMV) Control Slides contain sections of positive staining lung and negative staining myometrium.