Learn More
Sino Biological Human cytomegalovirus (HCMV) Glycoprotein B / gB Antibody (Biotin), Rabbit MAb
Supplier:  Sino Biological 10202R038B100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CMV gB (Catalog#10202-V08H1; AAA45920.1) extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907), and then biotinylated.
Catalog No. 50-255-5437