Learn More
Sino Biological CMV gB PAb 20ul (Sino Biological)
Supplier:  Sino Biological 10202RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human cytomegalovirus (HCMV) Glycoprotein B / gB (Catalog#10202-V08H1; AAA45920.1; Met1-Lys700 & Arg777-Val907). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3558