Learn More
Drg International Inc DRG CMV IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3468R
Encompass_Preferred
An enzyme immunoassay for the quantitative and qualitative determination of IgG-class antibodies to Cytomegalovirus (CMV) in human serum and plasma (EDTA-, heparin, or citrate plasma). This product is for Research Use Only. The DRG CMV IgG ELISA Kit is a solid phase enzyme linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with inactivated grade 2 Cytomegalovirus (CMV) antigen (strain AD-169).
Catalog No. 50-194-7565