Learn More
Drg International Inc DRG CMV IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3469R
Encompass_Preferred
An enzyme immunoassay for the quantitative and qualitative determination of IgM-class antibodies to Cytomegalovirus (CMV) in human serum and plasma (EDTA, heparin or citrate plasma). This product is for Research Use Only. The DRG Cytomegalovirus (CMV) IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent to eliminate competitive inhibition from specific IgG. Microtiter wells as a solid phase are coated with inactivated grade 2 Cytomegalovirus (CMV) antigen (strain AD-169). 
Catalog No. 50-194-7566