Learn More
Enzo Life Sciences CMV native antigen (100µg)
SDP
Supplier:  Enzo Life Sciences ALX6301250100
Encompass_Preferred
Alternative name: Cytomegalovirus native antigen. Purity: ≥95%. Appearance: Cloudy liquid. Formulation: Liquid. In 0.2% Triton STE. Source: From cytomegalovirus, strain AD169. Inactivated by 0.2% Triton X-100. Long Term Storage: -80°C. Applications: ELISA Positive control, Antibody capture.
Catalog No. 50-200-9651