Learn More
Mabtech Inc PepPool: CMV (CD4 and CD8), human, 25 μg of each peptide, lyophilized, 42 peptides, >95% purity, MHC class I: 28, MHC class II: 14
Catalog No. NC1916366
Encompass
PepPool: CMV (CD4 and CD8), human, 25 μg of each peptide, lyophilized, 42 peptides, >95% purity, MHC class I: 28, MHC class II: 14
Catalog No. NC1916366 Supplier Mabtech Inc Supplier No. 36191
PepPool: CMV (CD4 and CD8), human, 25 μg of each peptide, lyophilized, 42 peptides, >95% purity, MHC class I: 28, MHC class II: 14